8-Methylimidazo[1,2-a]pyridine-2-carboxylic acid 95%
Supplier: Apollo Scientific
OR912859-1G OR912859-250MG
APOSOR912859-1GEA 270 EUR
APOSOR912859-1G APOSOR912859-250MG
8-Methylimidazo[1,2-a]pyridine-2-carboxylic acid 95%
8-Methylimidazo[1,2-a]pyridine-2-carboxylic acid
Formula: C₉H₈N₂O₂
Storage Temperature: Ambient
CAS Number: 88751-05-7