trans-(1S,2S)-N,N'-Dimethylcyclohexane-1,2-diamine 97%
Supplier: Apollo Scientific
Synonyms: (1S,2S)-(+)-trans-1,2-Bis(methylamino)cyclohexane, (1S,2S)-N,N'-Dimethylcyclohexane-1,2-diamine
OR913416-10G OR913416-25G OR913416-5G
APOSOR913416-10GEA 50.9 EUR
APOSOR913416-10G APOSOR913416-25G APOSOR913416-5G
trans-(1S,2S)-N,N'-Dimethylcyclohexane-1,2-diamine 97%
trans-(1S,2S)-N,N'-Dimethylcyclohexane-1,2-diamine
Formula: C₈H₁₈N₂
MW: 142.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00671528
CAS Number: 87583-89-9