(S)-(+)-2,2-Dimethylcyclopropanecarboxylic acid 95%
Supplier: Apollo Scientific
Synonyms: (S)-2,2-Dimethylcyclopropanecarboxylic acid
OR913786-10G OR913786-1G OR913786-5G
APOSOR913786-10GEA 498 EUR
APOSOR913786-10G APOSOR913786-1G APOSOR913786-5G
(S)-(+)-2,2-Dimethylcyclopropanecarboxylic acid 95%
(S)-(+)-2,2-Dimethylcyclopropanecarboxylic acid
Formula: C₆H₁₀O₂
MW: 114.1400 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08460918
CAS Number: 14590-53-5