L-(+)-Pantolactone 97%
Supplier: Apollo Scientific
Synonyms: (S)-3-Hydroxy-4,4-dimethyldihydrofuran-2(3H)-one
OR913788-1G OR913788-5G
APOSOR913788-1GEA 63.8 EUR
APOSOR913788-1G APOSOR913788-5G
L-(+)-Pantolactone 97%
L-(+)-Pantolactone
Formula: C₆H₁₀O₃
MW: 130.143 g/mol
Boiling Pt: 120…122 °C
Melting Pt: 89…93 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00216625
CAS Number: 5405-40-3