3-(1H-1,2,4-Triazol-1-yl)aniline 90%
Supplier: Apollo Scientific
Warning
Synonyms: 3-(1,2,4-Triazol-1-yl)aniline
OR9139-1G OR9139-250MG
APOSOR9139-1GEA 151 EUR
APOSOR9139-1G APOSOR9139-250MG
3-(1H-1,2,4-Triazol-1-yl)aniline 90%
3-(1H-1,2,4-Triazol-1-yl)aniline
Formula: C₈H₈N₄
Melting Pt: 102…107 °C
Storage Temperature: Ambient
MDL Number: MFCD08271899
CAS Number: 176032-78-3
UN: 2811
ADR: 6.1,III