1,2,3,4-Tetrahydronaphthalen-2-amine hydrochloride 97%
Supplier: Apollo Scientific
OR914164-10G OR914164-1G OR914164-5G
APOSOR914164-10GEA 676 EUR
APOSOR914164-10G APOSOR914164-1G APOSOR914164-5G
1,2,3,4-Tetrahydronaphthalen-2-amine hydrochloride 97%
1,2,3,4-Tetrahydronaphthalen-2-amine hydrochloride
Formula: C₁₀H₁₄ClN
Storage Temperature: Ambient
CAS Number: 1743-01-7