o-Anisaldehyde 98%
Supplier: Apollo Scientific
Warning
Synonyms: Salicylaldehyde methyl ether, 2-Methoxybenzaldehyde, 2-Anisaldehyde
OR914416-100G OR914416-25G OR914416-5G
APOSOR914416-100GEA 29 EUR
APOSOR914416-100G APOSOR914416-25G APOSOR914416-5G
o-Anisaldehyde 98%
o-Anisaldehyde
Formula: C₈H₈O₂
Boiling Pt: 238 °C
Melting Pt: 34…40 °C
Flash Pt: 117 °C
Storage Temperature: Ambient
MDL Number: MFCD00003308
CAS Number: 135-02-4