3-Methyl-2-naphthol 98%
Supplier: Apollo Scientific
Synonyms: 2-Hydroxy-3-methylnaphthalene, 3-Methylnaphthalen-2-ol
OR914496-1G OR914496-5G
APOSOR914496-1GEA 252 EUR
APOSOR914496-1G APOSOR914496-5G
3-Methyl-2-naphthol 98%
3-Methyl-2-naphthol
Formula: C₁₁H₁₀O
MW: 158.1965 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09037754
CAS Number: 17324-04-8