1-methyl-1H-indole-5-carbaldehyde 97%
Supplier: Apollo Scientific
Warning
OR9157-1G OR9157-250MG OR9157-5G
APOSOR9157-1GEA 72.5 EUR
APOSOR9157-1G APOSOR9157-250MG APOSOR9157-5G
1-methyl-1H-indole-5-carbaldehyde 97%
1-methyl-1H-indole-5-carbaldehyde
Formula: C₁₀H₉NO
MW: 159.188 g/mol
Melting Pt: 84…86 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08271908
CAS Number: 90923-75-4
UN: 2811
ADR: 6.1 III