4-(Piperidin-1-ylmethyl)phenylboronic acid pinacol ester
Supplier: Apollo Scientific
Danger
Synonyms: 1-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)piperidine, 4-(1-Piperidinylmethyl)benzeneboronic acid pinacol ester
OR9166-1G OR9166-5G
APOSOR9166-1GEA 75.9 EUR
APOSOR9166-1G APOSOR9166-5G
4-(Piperidin-1-ylmethyl)phenylboronic acid pinacol ester
4-(Piperidin-1-ylmethyl)phenylboronic acid pinacol ester
Formula: C₁₈H₂₈BNO₂
MW: 301.23 g/mol
Melting Pt: 68…71 °C
Storage Temperature: Ambient
MDL Number: MFCD08271932
CAS Number: 859833-22-0
UN: 3259
ADR: 8 III