1,2-Indanedione 97%
Supplier: Apollo Scientific
Synonyms: 1H-Indene-1,2(3H)-dione, 1,2-Indandione
OR916829-10G OR916829-5G OR916829-25G
APOSOR916829-10GEA 477 EUR
APOSOR916829-10G APOSOR916829-5G APOSOR916829-25G
1,2-Indanedione 97%
1,2-Indanedione
Formula: C₉H₆O₂
MW: 146.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00463378
CAS Number: 16214-27-0