1-(4-Chlorophenyl)cyclopropanamine 95%
Supplier: Apollo Scientific
OR916952-1G OR916952-250MG
APOSOR916952-1GEA 142 EUR
APOSOR916952-1G APOSOR916952-250MG
1-(4-Chlorophenyl)cyclopropanamine 95%
1-(4-Chlorophenyl)cyclopropanamine
Formula: C₉H₁₀ClN
MW: 167.64 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07371527
CAS Number: 72934-36-2