3-(3-Bromophenyl)-5-methyl-1,2,4-oxadiazole 97%
Supplier: Apollo Scientific
Warning
OR9178-1G OR9178-25G OR9178-5G
APOSOR9178-1GEA 87.3 EUR
APOSOR9178-1G APOSOR9178-25G APOSOR9178-5G
3-(3-Bromophenyl)-5-methyl-1,2,4-oxadiazole 97%
3-(3-Bromophenyl)-5-methyl-1,2,4-oxadiazole
Formula: C₉H₇BrN₂O
Melting Pt: 81…83 °C
Storage Temperature: Ambient
MDL Number: MFCD08087563
CAS Number: 160377-57-1