4-Methyl-2-(2-methyl-1-propenyl)tetrahydropyran (cis- and trans- mixture) 95%
Supplier: Apollo Scientific
Synonyms: 4-Methyl-2-(2-methylpropenyl)tetrahydro-2H-pyran, 4-Methyl-2-(2-methylprop-1-en-1-yl)tetrahydro-2H-pyran
OR918409-25G OR918409-5G
APOSOR918409-25GEA 197 EUR
APOSOR918409-25G APOSOR918409-5G
4-Methyl-2-(2-methyl-1-propenyl)tetrahydropyran (cis- and trans- mixture) 95%
4-Methyl-2-(2-methyl-1-propenyl)tetrahydropyran (cis- and trans- mixture)
Formula: C₁₀H₁₈O
MW: 154.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00036607
CAS Number: 16409-43-1