1-(3-Aminophenyl)-3-methyl-2-imidazolidinone 95%
Supplier: Apollo Scientific
Synonyms: 1-(3-Aminophenyl)-3-methylimidazolidin-2-one
OR918500-1G OR918500-250MG
APOSOR918500-1GEA 200 EUR
APOSOR918500-1G APOSOR918500-250MG
1-(3-Aminophenyl)-3-methyl-2-imidazolidinone 95%
1-(3-Aminophenyl)-3-methyl-2-imidazolidinone
Formula: C₁₀H₁₃N₃O
MW: 191.2300 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05864562
CAS Number: 517918-82-0