1-(2-Hydroxyethyl)-4-(3-hydroxypropyl)piperidine 98%
Supplier: Apollo Scientific
Synonyms: 1-Ethanol-4-propanolpiperidine
OR919282-25G OR919282-5G
APOSOR919282-25GEA 306 EUR
APOSOR919282-25G APOSOR919282-5G
1-(2-Hydroxyethyl)-4-(3-hydroxypropyl)piperidine 98%
1-(2-Hydroxyethyl)-4-(3-hydroxypropyl)piperidine
Formula: C₁₀H₂₁NO₂
MW: 187.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00047481
CAS Number: 19780-85-9