Methyl-3,3-dimethyl-4-pentenoate 95%
Supplier: Apollo Scientific
Synonyms: Methyl 3,3-dimethylpent-4-enoate, 3,3-Dimethyl-4-pentenoic Acid Methyl Ester
OR919294-25G OR919294-5G
APOSOR919294-25GEA 232 EUR
APOSOR919294-25G APOSOR919294-5G
Methyl-3,3-dimethyl-4-pentenoate 95%
Methyl-3,3-dimethyl-4-pentenoate
Formula: C₈H₁₄O₂
MW: 142.20 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00075381
CAS Number: 63721-05-1