3-Hepten-2-one 96%
Supplier: Apollo Scientific
Synonyms: Hept-3-en-2-one
OR919591-25G OR919591-5G
APOSOR919591-25GEA 255 EUR
APOSOR919591-25G APOSOR919591-5G
3-Hepten-2-one 96%
3-Hepten-2-one
Formula: C₇H₁₂O
MW: 112.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00015564
CAS Number: 1119-44-4