2-Ethylcyclohexanol 95%

Supplier: Apollo Scientific

OR919772-25G
APOSOR919772-25GEA 184 EUR
APOSOR919772-25G
2-Ethylcyclohexanol 95%
2-Ethylcyclohexanol
Formula: C₈H₁₆O
MW: 128.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00001445
CAS Number: 3760-20-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR