cis-4-tert-Butylcyclohexanol 95%
Supplier: Apollo Scientific
Synonyms: cis-4-(tert-Butyl)cyclohexanol
OR920231-1G OR920231-250MG
APOSOR920231-1GEA 331 EUR
APOSOR920231-1G APOSOR920231-250MG
cis-4-tert-Butylcyclohexanol 95%
cis-4-tert-Butylcyclohexanol
Formula: C₁₀H₂₀O
MW: 156.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00070476
CAS Number: 937-05-3