N,N'-Diacetyl-1,4-phenylenediamine 95%
Supplier: Apollo Scientific
Synonyms: DAPPD, N,N'-1,4-Phenylenebisacetamide, N,N'-(1,4-Phenylene)diacetamide
OR920370-25G
APOSOR920370-25GEA 172 EUR
APOSOR920370-25G
N,N'-Diacetyl-1,4-phenylenediamine 95%
N,N'-Diacetyl-1,4-phenylenediamine
Formula: C₁₀H₁₂N₂O₂
MW: 192.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00026142
CAS Number: 140-50-1