3-Cyclohexyl-1-propanol 95%
Supplier: Apollo Scientific
Synonyms: 3-Hydroxypropylcyclohexane, 3-Cyclohexylpropan-1-ol
OR920665-25G OR920665-5G
APOSOR920665-25GEA 335 EUR
APOSOR920665-25G APOSOR920665-5G
3-Cyclohexyl-1-propanol 95%
3-Cyclohexyl-1-propanol
Formula: C₉H₁₈O
MW: 142.24 g/mol
Boiling Pt: 223
Melting Pt: -60
Storage Temperature: Ambient
MDL Number: MFCD00001528
CAS Number: 1124-63-6