2,4,6-Trimethoxypyrimidine 95%
Supplier: Apollo Scientific
OR920795-1G
OR920795-5G
APOSOR920795-1GEA
36.4
EUR
APOSOR920795-1G
APOSOR920795-5G
2,4,6-Trimethoxypyrimidine 95%
2,4,6-Trimethoxypyrimidine
Formula:
C₇H₁₀N₂O₃ MW: 170.17 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00829225 CAS Number: 13106-85-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...