1H-Pyrazolo[4,3-c]pyridine-3-carboxylic acid 95%
Supplier: Apollo Scientific
OR921183-100MG OR921183-1G OR921183-250MG
APOSOR921183-100MGEA 161 EUR
APOSOR921183-100MG APOSOR921183-1G APOSOR921183-250MG
1H-Pyrazolo[4,3-c]pyridine-3-carboxylic acid 95%
1H-Pyrazolo[4,3-c]pyridine-3-carboxylic acid
Formula: C₇H₅N₃O₂
MW: 163.1335 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11042720
CAS Number: 932702-11-9