4-Chloro-5-methyl-o-anisidine 95%
Supplier: Apollo Scientific
Synonyms: 4-Chloro-2-methoxy-5-methylaniline
OR921813-1G OR921813-5G
APOSOR921813-1GEA 125 EUR
APOSOR921813-1G APOSOR921813-5G
4-Chloro-5-methyl-o-anisidine 95%
4-Chloro-5-methyl-o-anisidine
Formula: C₈H₁₀ClNO
Storage Temperature: Ambient
CAS Number: 6376-14-3