1,3,5-Trimethyl-1H-pyrazole 98%
Supplier: Apollo Scientific
Danger
Synonyms: 1,3,5-Trimethyl-1H-pyrazole, 1,3,5-Trimethylpyrazole
OR9220-25G OR9220-5G OR9220-1G
APOSOR9220-25GEA 246 EUR
APOSOR9220-25G APOSOR9220-5G APOSOR9220-1G
1,3,5-Trimethyl-1H-pyrazole 98%
1,3,5-Trimethyl-1H-pyrazole
Formula: C₆H₁₀N₂
MW: 110.16 g/mol
Boiling Pt: 170 °C
Melting Pt: 37 °C
Storage Temperature: Ambient
MDL Number: MFCD00015536
CAS Number: 1072-91-9
UN: 1325
ADR: 4.1,III