6-Methoxybenzo[1,3]dioxole-5-carbaldehyde 95%
Supplier: Apollo Scientific
OR922248-1G OR922248-5G
APOSOR922248-1GEA 161 EUR
APOSOR922248-1G APOSOR922248-5G
6-Methoxybenzo[1,3]dioxole-5-carbaldehyde 95%
6-Methoxybenzo[1,3]dioxole-5-carbaldehyde
Formula: C₉H₈O₄
MW: 180.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00598413
CAS Number: 5780-00-7