2,3,5-Trimethylhydroquinone 95%

Supplier: Apollo Scientific
Danger

APOSOR922335-100GEA 125 EUR
APOSOR922335-100G
2,3,5-Trimethylhydroquinone 95%
2,3,5-Trimethylhydroquinone
Formula: C₉H₁₂O₂
MW: 152,19 g/mol
Boiling Pt: 295 °C/760mmHg
Melting Pt: 169 °C
Flash Pt: 191 °C
Storage Temperature: Ambient
MDL Number: MFCD00002346
UN: 3077
ADR: 9+EHS,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR