2,2-Dimethyl-1,3-benzodioxole 95%
Supplier: Apollo Scientific
Synonyms: 1,2-(Isopropylidenedioxy)benzene, 2,2-Dimethylbenzo[d][1,3]dioxole
OR922649-25G OR922649-5G
APOSOR922649-25GEA 179 EUR
APOSOR922649-25G APOSOR922649-5G
2,2-Dimethyl-1,3-benzodioxole 95%
2,2-Dimethyl-1,3-benzodioxole
Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08061594
CAS Number: 14005-14-2