Methyl-4-hydroxybenzoate 98%
Supplier: Apollo Scientific
Synonyms: Methylparaben, 4-Hydroxybenzoic acid methyl ester, Methyl Parahydroxybenzoate, Methyl 4-hydroxybenzoate
OR922886-100G OR922886-1KG OR922886-500G
APOSOR922886-100GEA 21.9 EUR
APOSOR922886-100G APOSOR922886-1KG APOSOR922886-500G
Methyl-4-hydroxybenzoate 98%
Methyl 4-hydroxybenzoate
Formula: HOC₆H₄CO₂CH₃
MW: 152.15 g/mol
Melting Pt: 125…128 °C
Storage Temperature: Ambient
MDL Number: MFCD00002352
CAS Number: 99-76-3
Merck Index: 13,06129