Ethyl-3-cyclohexene-1-carboxylate 95%
Supplier: Apollo Scientific
Synonyms: 3-Cyclohexene-1-carboxylic Acid Ethyl Ester
OR922945-10G OR922945-1G OR922945-5G
APOSOR922945-10GEA 328 EUR
APOSOR922945-10G APOSOR922945-1G APOSOR922945-5G
Ethyl-3-cyclohexene-1-carboxylate 95%
Ethyl-3-cyclohexene-1-carboxylate
Formula: C₉H₁₄O₂
MW: 154.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00037158
CAS Number: 15111-56-5