Diisopropylamine hydrochloride 99%
Supplier: Apollo Scientific
Warning
OR923049-25G OR923049-5G OR923049-500G
APOSOR923049-25GEA 109 EUR
APOSOR923049-25G APOSOR923049-5G APOSOR923049-500G
Diisopropylamine hydrochloride 99%
Diisopropylamine hydrochloride
Formula: C₆H₁₅N
MW: 137.65 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00034860
CAS Number: 819-79-4