Ethyl propionate 95%
Supplier: Apollo Scientific
OR923289-100G OR923289-1KG OR923289-500G
APOSOR923289-100GEA 125 EUR
APOSOR923289-100G APOSOR923289-1KG APOSOR923289-500G
Ethyl propionate 95%
Ethyl propionate
Formula: C₅H₁₀O₂
Storage Temperature: Ambient
MDL Number: MFCD00009308
CAS Number: 105-37-3