(R)-(+)-4-(Methoxymethyl)-1,3-dioxolan-2-one 95%
Supplier: Apollo Scientific
Synonyms: (R)-(+)-3-Methoxypropylene Carbonate, (R)-(+)-4-(Methoxymethyl)-2-oxo-1,3-dioxolane
OR923363-1G OR923363-25G OR923363-5G
APOSOR923363-1GEA 132 EUR
APOSOR923363-1G APOSOR923363-25G APOSOR923363-5G
(R)-(+)-4-(Methoxymethyl)-1,3-dioxolan-2-one 95%
(R)-(+)-4-(Methoxymethyl)-1,3-dioxolan-2-one
Formula: C₅H₈O₄
MW: 132.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01321303
CAS Number: 185836-34-4