4-tert-Butylcyclohexanone 98%
Supplier: Apollo Scientific
OR923989-100G OR923989-500G
APOSOR923989-100GEA 59.6 EUR
APOSOR923989-100G APOSOR923989-500G
4-tert-Butylcyclohexanone 98%
4-tert-Butylcyclohexanone
Formula: C₁₀H₁₈O
MW: 154.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00001642
CAS Number: 98-53-3