tert-Butyl-1-oxo-2,8-diazaspiro[4.5]decane-8-carboxylate 97%
Supplier: Apollo Scientific
Warning
Synonyms: tert-Butyl 1-oxo-2,8-diazaspiro[4.5]decane-8-carboxylate, 2,8-Diazaspiro[4.5]decan-1-one, N8-BOC protected
OR924090-5G
APOSOR924090-5GEA 192 EUR
APOSOR924090-5G
tert-Butyl-1-oxo-2,8-diazaspiro[4.5]decane-8-carboxylate 97%
tert-Butyl-1-oxo-2,8-diazaspiro[4.5]decane-8-carboxylate
Formula: C₁₃H₂₂N₂O₃
MW: 254.33 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08461248
CAS Number: 268550-48-7