4-Hydroxy-4-methyl-2-pentanone 95%
Supplier: Apollo Scientific
Synonyms: Diacetone alcohol
OR924093-100G OR924093-1KG OR924093-500G
APOSOR924093-100GEA 125 EUR
APOSOR924093-100G APOSOR924093-1KG APOSOR924093-500G
4-Hydroxy-4-methyl-2-pentanone 95%
4-Hydroxy-4-methyl-2-pentanone
Formula: (CH₃)₂C(OH)CH₂COCH₃
MW: 116.16 g/mol
Boiling Pt: 166
Melting Pt: -44
Flash Pt: 58
Storage Temperature: Ambient
MDL Number: MFCD00004471
CAS Number: 123-42-2
Merck Index: 13,02983