(S)-(-)-4-(Methoxymethyl)-1,3-dioxolan-2-one 95%
Supplier: Apollo Scientific
Synonyms: (S)-(-)-3-Methoxypropylene Carbonate, (S)-(-)-4-(Methoxymethyl)-2-oxo-1,3-dioxolane
OR924119-1G OR924119-25G OR924119-5G
APOSOR924119-1GEA 132 EUR
APOSOR924119-1G APOSOR924119-25G APOSOR924119-5G
(S)-(-)-4-(Methoxymethyl)-1,3-dioxolan-2-one 95%
(S)-(-)-4-(Methoxymethyl)-1,3-dioxolan-2-one
Formula: C₅H₈O₄
MW: 132.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01321302
CAS Number: 135682-18-7