2,4-Dimethyl-1,3-dioxane 95%
Supplier: Apollo Scientific
OR924236-10G OR924236-1G OR924236-5G
APOSOR924236-10GEA 270 EUR
APOSOR924236-10G APOSOR924236-1G APOSOR924236-5G
2,4-Dimethyl-1,3-dioxane 95%
2,4-Dimethyl-1,3-dioxane
Formula: C₆H₁₂O₂
MW: 116.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00023821
CAS Number: 766-20-1