4-Chloro-N-methylaniline 98%
Supplier: Apollo Scientific
OR924273-100G
OR924273-25G
APOSOR924273-100GEA
317
EUR
APOSOR924273-100G
APOSOR924273-25G
4-Chloro-N-methylaniline 98%
4-Chloro-N-methylaniline
Formula:
C₇H₈ClN MW: 141.6 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00000614 CAS Number: 932-96-7 |