3,3-Dimethoxypropiononitrile 93%
Supplier: Apollo Scientific
Synonyms: 3,3-Dimethoxypropanenitrile
OR924380-5G OR924380-1G OR924380-10G
APOSOR924380-5GEA 287 EUR
APOSOR924380-5G APOSOR924380-1G APOSOR924380-10G
3,3-Dimethoxypropiononitrile 93%
3,3-Dimethoxypropiononitrile
Formula: C₅H₉NO₂
MW: 115.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00044797
CAS Number: 57597-62-3