tert-Butyl-2-(tetrahydro-4H-pyran-4-ylidene)acetate 95%
Supplier: Apollo Scientific
OR924830-1G OR924830-250MG
APOSOR924830-1GEA 806 EUR
APOSOR924830-1G APOSOR924830-250MG
tert-Butyl-2-(tetrahydro-4H-pyran-4-ylidene)acetate 95%
tert-Butyl-2-(tetrahydro-4H-pyran-4-ylidene)acetate
Formula: C₁₁H₁₈O₃
MW: 198.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22415212
CAS Number: 894789-82-3