3,4-Dimethylcyclohexanol (mixture of isomers) 95%
Supplier: Apollo Scientific
Synonyms: 3,4-Dimethylcyclohexanol
OR925006-5G
APOSOR925006-5GEA 60.9 EUR
APOSOR925006-5G
3,4-Dimethylcyclohexanol (mixture of isomers) 95%
3,4-Dimethylcyclohexanol (mixture of isomers)
Formula: C₈H₁₆O
MW: 128.212 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00001506
CAS Number: 5715-23-1