8-Nitroquinaldine 98%

Supplier: Apollo Scientific

OR925095-25G OR925095-5G
APOSOR925095-25GEA 122 EUR
APOSOR925095-25G APOSOR925095-5G
8-Nitroquinaldine 98%
8-Nitroquinaldine
Formula: C₁₀H₈N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00006764
CAS Number: 881-07-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR