Phenylphosphonic acid 95%
Supplier: Apollo Scientific
Synonyms:
Phenylphosphonic Acid
OR925125-500G
APOSOR925125-500GEA
78.3
EUR
APOSOR925125-500G
Phenylphosphonic acid 95%
Phenylphosphonic acid
Formula:
C₆H₇O₃P MW: 158.09 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00002136 CAS Number: 1571-33-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...