4,4'-Bipiperidine 98%
Supplier: Apollo Scientific
OR925367-1G OR925367-5G
APOSOR925367-1GEA 179 EUR
APOSOR925367-1G APOSOR925367-5G
4,4'-Bipiperidine 98%
4,4'-Bipiperidine
Formula: C₁₀H₂₀N₂
MW: 168.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01104315
CAS Number: 15336-72-8