3-(Chloromethyl)-5-isopropyl-1,2,4-oxadiazole 95%
Supplier: Apollo Scientific
OR925985-1G OR925985-250MG
APOSOR925985-1GEA 806 EUR
APOSOR925985-1G APOSOR925985-250MG
3-(Chloromethyl)-5-isopropyl-1,2,4-oxadiazole 95%
3-(Chloromethyl)-5-isopropyl-1,2,4-oxadiazole
Formula: C₆H₉ClN₂O
MW: 160.6 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09864313
CAS Number: 189130-87-8