1-[2-(1H-1,2,4-Triazol-1-yl)phenyl]methanamine hydrochloride 95%
Supplier: Apollo Scientific
Synonyms: 2-(1H-1,2,4-Triazol-1-yl)benzenemethanamine
OR926379-1G OR926379-250MG
APOSOR926379-1GEA 864 EUR
APOSOR926379-1G APOSOR926379-250MG
1-[2-(1H-1,2,4-Triazol-1-yl)phenyl]methanamine hydrochloride 95%
1-[2-(1H-1,2,4-Triazol-1-yl)phenyl]methanamine hydrochloride
Formula: C₉H₁₁ClN₄
MW: 210.67 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12027080
CAS Number: 449756-97-2