3-(Methylthio)benzoic acid 98+%
Supplier: Apollo Scientific
Warning
OR9264-1G OR9264-25G OR9264-5G
APOSOR9264-1GEA 21.9 EUR
APOSOR9264-1G APOSOR9264-25G APOSOR9264-5G
3-(Methylthio)benzoic acid 98+%
3-(Methylthio)benzoic acid
Formula: C₈H₈O₂S
MW: 168.22 g/mol
Melting Pt: 126…130 °C
Storage Temperature: Ambient
MDL Number: MFCD00156993
CAS Number: 825-99-0