Thiophene-2-carboxaldehyde diethylacetal 95%
Supplier: Apollo Scientific
Synonyms: 2-(Diethoxymethyl)thiophene
OR926523-1G OR926523-5G
APOSOR926523-1GEA 125 EUR
APOSOR926523-1G APOSOR926523-5G
Thiophene-2-carboxaldehyde diethylacetal 95%
Thiophene-2-carboxaldehyde diethylacetal
Formula: C₉H₁₄O₂S
MW: 186.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11109382
CAS Number: 13959-97-2